AMBH58176633 Ambeed, Inc.

2-Methoxyphenothiazine

  • MDL number MFCD00037838
  • Cas Number : 1771-18-2
  • Molecular Weight : 229.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1771-18-2.html

Safety Information

signalword Danger
pcodes P261-P273-P305+P351+P338
MSDS https://www.ambeed.com/products/1771-18-2.html
pictograms GHS07,GHS09
hstatements H315-H319-H335-H411
Availability details for
Quantity
  • Availability
  • Add to Cart