AMBH5817663A Ambeed, Inc.

2-Bromo-4-methylthiazole-5-carbonitrile

  • MDL number MFCD30292968
  • Cas Number : 1254034-55-3
  • Molecular Weight : 203.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1254034-55-3.html

Safety Information

MSDS https://www.ambeed.com/products/1254034-55-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart