AMBH5817663E Ambeed, Inc.

6-Chloro-N-methyl-5-nitro-4-pyrimidinamine

  • MDL number MFCD00209251
  • Cas Number : 23126-82-1
  • Molecular Weight : 188.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/23126-82-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/23126-82-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart