AMBH58176653 Ambeed, Inc.

2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-chlorobenzoic acid

  • MDL number MFCD02682157
  • Cas Number : 332121-92-3
  • Molecular Weight : 393.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/332121-92-3.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/332121-92-3.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart