AMBH58176660 Ambeed, Inc.

Fmoc-Arg(Mtr)-OH

  • MDL number MFCD00043098
  • Cas Number : 98930-01-9
  • Molecular Weight : 608.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/98930-01-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/98930-01-9.html
pictograms GHS07
hstatements H302+H312+H332-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart