AMBH5817667A Ambeed, Inc.

3-Chloro-2-fluoro-5-(trifluoromethyl)phenol

  • MDL number MFCD01631514
  • Cas Number : 261763-12-6
  • Molecular Weight : 214.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/261763-12-6.html

Safety Information

MSDS https://www.ambeed.com/products/261763-12-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart