AMBH5817667C Ambeed, Inc.

Sodium 2-iodobenzenesulfonate

  • MDL number MFCD12031329
  • Cas Number : 62973-69-7
  • Molecular Weight : 306.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/62973-69-7.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
MSDS https://www.ambeed.com/products/62973-69-7.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart