AMBH58176683 Ambeed, Inc.

tert-Butyl 3-chloro-7,8-dihydro-1,6-naphthyridine-6(5H)-carboxylate

  • MDL number MFCD16660015
  • Cas Number : 625099-34-5
  • Molecular Weight : 268.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/625099-34-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/625099-34-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart