AMBH5817668A Ambeed, Inc.

(S)-5-(Allyloxy)-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid

  • MDL number MFCD00273402
  • Cas Number : 132286-79-4
  • Molecular Weight : 287.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/132286-79-4.html

Safety Information

MSDS https://www.ambeed.com/products/132286-79-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart