AMBH5817668E Ambeed, Inc.

1,1-Di-n-octyl-4,4-bipyridinium Dibromide

  • MDL number MFCD00060051
  • Cas Number : 36437-30-6
  • Molecular Weight : 542.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,1'-Dioctyl-[4,4'-bipyridine]-1,1'-diium bromide
Purity 95
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/36437-30-6.html

Safety Information

signalword Warning
pcodes P261-P301+P312-P302+P352-P304+P340-P305+P351+P338
MSDS https://www.ambeed.com/products/36437-30-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart