AMBH58176698 Ambeed, Inc.

Dimethyl dodecanedioate

  • MDL number MFCD00043632
  • Cas Number : 1731-79-9
  • Molecular Weight : 258.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1731-79-9.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/1731-79-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart