AMBH581766A2 Ambeed, Inc.

tert-Butyl 1-chloro-2-oxo-6,9,12,15-tetraoxa-3-azaoctadecan-18-oate

  • MDL number MFCD22056319
  • Cas Number : 1365655-90-8
  • Molecular Weight : 397.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1365655-90-8.html

Safety Information

MSDS https://www.ambeed.com/products/1365655-90-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart