AMBH581766AC Ambeed, Inc.

(E)-Methyl 2-(methoxyimino)-2-(o-tolyl)acetate

  • MDL number MFCD16038452
  • Cas Number : 120974-97-2
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/120974-97-2.html

Safety Information

MSDS https://www.ambeed.com/products/120974-97-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart