AMBH581766B0 Ambeed, Inc.

(2R,5S)-1-(tert-Butoxycarbonyl)-5-phenylpyrrolidine-2-carboxylic acid

  • MDL number MFCD01321009
  • Cas Number : 158706-46-8
  • Molecular Weight : 291.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/158706-46-8.html

Safety Information

MSDS https://www.ambeed.com/products/158706-46-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart