AMBH581766B2 Ambeed, Inc.

Ethyl 3-(2,6-dichlorophenyl)-3-oxopropanoate

  • MDL number MFCD08458321
  • Cas Number : 72835-87-1
  • Molecular Weight : 261.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/72835-87-1.html

Safety Information

MSDS https://www.ambeed.com/products/72835-87-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart