AMBH581766C3 Ambeed, Inc.

1-(4-Fluoronaphthalen-1-yl)ethanone

  • MDL number MFCD00134475
  • Cas Number : 316-68-7
  • Molecular Weight : 188.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/316-68-7.html

Safety Information

signalword Warning
pcodes P261-P301+P312-P302+P352-P304+P340-P305+P351+P338
MSDS https://www.ambeed.com/products/316-68-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart