AMBH581766C4 Ambeed, Inc.

Hexaketocyclohexane hydrate

  • MDL number MFCD00149075
  • Cas Number : 1894538-97-6
  • Molecular Weight : 312.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1894538-97-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1894538-97-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart