AMBH581766CA Ambeed, Inc.

N,N-Dimethylcapramide

  • MDL number MFCD00043725
  • Cas Number : 14433-76-2
  • Molecular Weight : 199.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/14433-76-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/14433-76-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart