AMBH581766CF Ambeed, Inc.

GPR84 Antagonist 8

  • MDL number N/A
  • Cas Number : 1445846-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1445846-30-9.html

Safety Information

MSDS https://www.ambeed.com/products/1445846-30-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart