AMBH581766D5 Ambeed, Inc.

2-(3-Phenoxyphenyl)acetaldehyde

  • MDL number MFCD10566816
  • Cas Number : 109330-01-0
  • Molecular Weight : 212.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/109330-01-0.html

Safety Information

MSDS https://www.ambeed.com/products/109330-01-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart