AMBH581766D7 Ambeed, Inc.

Sodium 2-amino-4-sulfobenzenesulfonate

  • MDL number MFCD00078393
  • Cas Number : 24605-36-5
  • Molecular Weight : 275.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/24605-36-5.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
MSDS https://www.ambeed.com/products/24605-36-5.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart