AMBH581766DD Ambeed, Inc.

(S)-4-Benzyl-3-(but-2-enoyl)oxazolidin-2-one

  • MDL number MFCD27937442
  • Cas Number : 133812-16-5
  • Molecular Weight : 245.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/133812-16-5.html

Safety Information

MSDS https://www.ambeed.com/products/133812-16-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart