AMBH581766DF Ambeed, Inc.

1-((Chloromethyl)sulfonyl)-4-methylbenzene

  • MDL number MFCD00134281
  • Cas Number : 7569-26-8
  • Molecular Weight : 204.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/7569-26-8.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
MSDS https://www.ambeed.com/products/7569-26-8.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart