AMBH581766E2 Ambeed, Inc.

2-Chloro-6-nitro-1H-benzo[d]imidazole

  • MDL number MFCD00769824
  • Cas Number : 5955-72-6
  • Molecular Weight : 197.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225-227
Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/5955-72-6.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/5955-72-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart