AMBH581766F8 Ambeed, Inc.

2-(Pyridin-2-ylsulfonyl)acetonitrile

  • MDL number MFCD00041443
  • Cas Number : 170449-34-0
  • Molecular Weight : 182.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 220.4°C
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 440.8°C at 760 mmHg
Product Link https://www.ambeed.com/products/170449-34-0.html

Safety Information

signalword Warning
pcodes P261-P264-P270-P271-P280-P302+P352-P304+P340-P312-P330-P363-P403-P501
MSDS https://www.ambeed.com/products/170449-34-0.html
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart