AMBH58176701 Ambeed, Inc.

(S)-2-(2-Oxopyrrolidin-1-yl)butanoic acid

  • MDL number MFCD08272027
  • Cas Number : 102849-49-0
  • Molecular Weight : 171.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/102849-49-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/102849-49-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart