AMBH5817670E Ambeed, Inc.

4-Methylpyrimidine-2(1H)-thione hydrochloride

  • MDL number MFCD00012783
  • Cas Number : 6959-66-6
  • Molecular Weight : 162.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/6959-66-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/6959-66-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart