AMBH58176710 Ambeed, Inc.

(S)-1H-Indol-3-yl 2-(4-methylphenylsulfonamido)propanoate

  • MDL number MFCD03701236
  • Cas Number : 75062-54-3
  • Molecular Weight : 358.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/75062-54-3.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/75062-54-3.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart