AMBH58176724 Ambeed, Inc.

6-(tert-Butyl)-3,4-dihydroisoquinolin-1(2H)-one

  • MDL number MFCD19441516
  • Cas Number : 1178884-68-8
  • Molecular Weight : 203.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1178884-68-8.html

Safety Information

MSDS https://www.ambeed.com/products/1178884-68-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart