AMBH58176728 Ambeed, Inc.

Dasatinib hydrochloride

  • MDL number MFCD26960525
  • Cas Number : 854001-07-3
  • Molecular Weight : 524.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/854001-07-3.html

Safety Information

MSDS https://www.ambeed.com/products/854001-07-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart