AMBH5817673D Ambeed, Inc.

6-(Trifluoromethyl)-1H-indazol-5-amine

  • MDL number MFCD24642227
  • Cas Number : 1000373-75-0
  • Molecular Weight : 201.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/1000373-75-0.html

Safety Information

MSDS https://www.ambeed.com/products/1000373-75-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart