AMBH58176757 Ambeed, Inc.

3-Chlorophenylthiourea

  • MDL number MFCD00022165
  • Cas Number : 4947-89-1
  • Molecular Weight : 186.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138°C at N/A mmHg
Synonym 1-(3-Chlorophenyl)thiourea
Purity 98
Flash Point 133.2°C
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 296.7°C at 760 mmHg
Product Link https://www.ambeed.com/products/4947-89-1.html

Safety Information

signalword Danger
pcodes P264-P270-P301+P310+P330-P405-P501
MSDS https://www.ambeed.com/products/4947-89-1.html
pictograms GHS06
hstatements H300
Availability details for
Quantity
  • Availability
  • Add to Cart