AMBH58176978 Ambeed, Inc.

Tris(4-nonylphenyl) phosphite

  • MDL number MFCD00015301
  • Cas Number : 3050-88-2
  • Molecular Weight : 689.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 110
Storage temp. Sealed in dry,Room Temperature
Physical Form Solid or Semi-solid or liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/3050-88-2.html

Safety Information

signalword Danger
pcodes P201-P202-P261-P264-P272-P273-P280-P281-P302+P352-P305+P351+P338+P310-P308+P313-P333+P313-P362-P391-P405-P501
hcodes 9
MSDS https://www.ambeed.com/products/3050-88-2.html
pictograms GHS05,GHS07,GHS08,GHS09
hstatements H315-H317-H318-H361-H400-H410
Availability details for
Quantity
  • Availability
  • Add to Cart