AMBH58176A1A Ambeed, Inc.

5-(4-Aminophenyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD00806252
  • Cas Number : 32058-82-5
  • Molecular Weight : 193.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Product Link https://www.ambeed.com/products/32058-82-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/32058-82-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart