AMBH58176A65 Ambeed, Inc.

1,3-Benzodioxole-4-methanamine

  • MDL number MFCD11858181
  • Cas Number : 182634-34-0
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Benzo[d][1,3]dioxol-4-ylmethanamine
Purity 95
Flash Point 124.5
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 266.6±19.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/182634-34-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/182634-34-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart