AMBH58176B3B Ambeed, Inc.

(2,3-Dihydrobenzofuran-5-yl)methanamine

  • MDL number MFCD00728869
  • Cas Number : 55745-74-9
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/55745-74-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/55745-74-9.html
pictograms GHS07
hstatements H302-H312-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart