AMBH6FD84616 Ambeed, Inc.

Methyl4-mercaptobenzoate

  • MDL number MFCD01863423
  • Cas Number : 6302-65-4
  • Molecular Weight : 168.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 33-36°C at N/A mmHg
Purity 95
Flash Point 167.9°C
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 266.7°C at 760 mmHg
Product Link https://www.ambeed.com/products/6302-65-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/6302-65-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart