AMBH93D52F93 Ambeed, Inc.

(3R,4S)-rel-4-(3-Bromophenyl)-1-(tert-butoxycarbonyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD06659267
  • Cas Number : 1161787-83-2
  • Molecular Weight : 370.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1161787-83-2.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/1161787-83-2.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart