AMBH93D52FAC Ambeed, Inc.

N1-Dodecylpropane-1,3-diamine

  • MDL number MFCD00014828
  • Cas Number : 5538-95-4
  • Molecular Weight : 242.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 326°C at 760 mmHg
Product Link https://www.ambeed.com/products/5538-95-4.html

Safety Information

signalword Danger
pcodes P260-P264-P270-P273-P280-P301+P312-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P321-P363-P391-P405-P501
MSDS https://www.ambeed.com/products/5538-95-4.html
pictograms GHS05,GHS07,GHS09
hstatements H302-H314-H400
Availability details for
Quantity
  • Availability
  • Add to Cart