AMBH93D52FB5 Ambeed, Inc.

Z-DEVD-FMK

  • MDL number MFCD23380217
  • Cas Number : 210344-95-9
  • Molecular Weight : 668.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (5S,8S,11S,14S)-methyl 14-(2-fluoroacetyl)-11-isopropyl-5-(2-methoxy-2-oxoethyl)-8-(3-methoxy-3-oxopropyl)-3,6,9,12-tetraoxo-1-phenyl-2-oxa-4,7,10,13-tetraazahexadecan-16-oate
Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/210344-95-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/210344-95-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart