AMBH93D52FBB Ambeed, Inc.

H-His(Trt)-OH

  • MDL number MFCD00153444
  • Cas Number : 35146-32-8
  • Molecular Weight : 397.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210.5°C
Synonym (S)-2-Amino-3-(1-trityl-1H-imidazol-4-yl)propanoic acid
Purity 97
Storage temp. Sealed in dry,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 584.8°C at 760 mmHg
Product Link https://www.ambeed.com/products/35146-32-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/35146-32-8.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart