AMBH93D52FD3 Ambeed, Inc.

(R)-2-Aminobutane-1,4-diol

  • MDL number MFCD18205987
  • Cas Number : 80572-06-1
  • Molecular Weight : 105.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form Yellow to Brown Sticky Oil to Solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/80572-06-1.html

Safety Information

signalword Danger
pcodes P260-P264-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/80572-06-1.html
pictograms GHS05
hstatements H314-H335
Availability details for
Quantity
  • Availability
  • Add to Cart