AMBH93D53050 Ambeed, Inc.

Zofenopril Calcium

  • MDL number MFCD08704648
  • Cas Number : 81938-43-4
  • Molecular Weight : 897.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 250�t N/A mmHg
bp 646.3�t 760 mmHg
Iupac Name calcium bis[(2S,4S)-1-[(2S)-3-(benzoylsulfanyl)-2-methylpropanoyl]-4-(phenylsulfanyl)-2-pyrrolidinecarboxylate]
Inchi Code 1S/2C22H23NO4S2.Ca/c2*1-15(14-28-22(27)16-8-4-2-5-9-16)20(24)23-13-18(12-19(23)21(25)26)29-17-10-6-3-7-11-17;/h2*2-11,15,18-19H,12-14H2,1H3,(H,25,26);/q;;+2/p-2/t2*15-,18+,19+;/m11./s1
Inchi Key NSYUKKYYVFVMST-LETVYOFWSA-L

Description

Melting Point 250�t N/A mmHg
Synonym Calcium (2S,4S)-1-((S)-3-(benzoylthio)-2-methylpropanoyl)-4-(phenylthio)pyrrolidine-2-carboxylate
Purity 98%
Flash Point 344.7
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 646.3�t 760 mmHg
Product Link https://www.ambeed.com/products/81938-43-4.html

Safety Information

signalword Danger
pcodes P273-P501
MSDS https://www.ambeed.com/products/81938-43-4.html
pictograms GHS09
hstatements H410
Availability details for
Quantity
  • Availability
  • Add to Cart