AMBH93D530B3 Ambeed, Inc.

Balovaptan

  • MDL number N/A
  • Cas Number : 1228088-30-9
  • Molecular Weight : 409.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 8-Chloro-5-methyl-1-((1r,4r)-4-(pyridin-2-yloxy)cyclohexyl)-5,6-dihydro-4H-benzo[f][1,2,4]triazolo[4,3-a][1,4]diazepine
Purity 97
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1228088-30-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1228088-30-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart