AMBH93D53676 Ambeed, Inc.

Terephthalic acid monoamide

  • MDL number MFCD00025482
  • Cas Number : 6051-43-0
  • Molecular Weight : 165.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 248°C at N/A mmHg
Purity 97
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 400.5°C at 760 mmHg
Product Link https://www.ambeed.com/products/6051-43-0.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
MSDS https://www.ambeed.com/products/6051-43-0.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart