AMBH93D5367D Ambeed, Inc.

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol

  • MDL number MFCD29920248
  • Cas Number : 2043962-01-0
  • Molecular Weight : 270.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/2043962-01-0.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/2043962-01-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart