AMBH93D53684 Ambeed, Inc.

2-(Methylthio)thiazole-4-carboxylic acid

  • MDL number MFCD11054031
  • Cas Number : 907543-75-3
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/907543-75-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/907543-75-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart