AMBH93D53686 Ambeed, Inc.

2-Phenyl-2-(pyridin-2-yl)acetonitrile

  • MDL number MFCD00023497
  • Cas Number : 5005-36-7
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87-89°C
Purity 97
Flash Point 115.2°C
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 322.3°C at 760 mmHg
Product Link https://www.ambeed.com/products/5005-36-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/5005-36-7.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart