AMBH93D5368C Ambeed, Inc.

tert-Butyl 3-aminoazepane-1-carboxylate

  • MDL number MFCD06738754
  • Cas Number : 609789-17-5
  • Molecular Weight : 214.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form Colorless to Pale-yellow to Yellow-brown Liquid or Semi-solid or Solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/609789-17-5.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/609789-17-5.html
pictograms GHS05
hstatements H314-H318
Availability details for
Quantity
  • Availability
  • Add to Cart