AMBH93D5368F Ambeed, Inc.

(S)-N-Methyl-1-(1-methylpyrrolidin-2-yl)methanamine

  • MDL number MFCD18829960
  • Cas Number : 175417-85-3
  • Molecular Weight : 128.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/175417-85-3.html

Safety Information

signalword Danger
pcodes P210-P261-P305+P351+P338
MSDS https://www.ambeed.com/products/175417-85-3.html
pictograms GHS02,GHS07
hstatements H225-H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart